Learn More
Epredia™ Tray 75mm x 75mm Epredia™ Microm
Brand:  Epredia™ 715400
Product Code. 12093139
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Tray
75 mm
75 mm
Product Title
Epredia™ Tray 75mm x 75mm Epredia™ Microm >