Learn More
VITLAB™ Transparent Polypropylene Watch Glasses
Brand: VITLAB™ 80455
This item is not returnable.
View return policy
Specifications
Autoclavable | |
Round | |
Used as Beaker Covers |
Polypropylene | |
Watch Glass | |
100 mm |
Product Content Correction
Product Title
VITLAB™ Transparent Polypropylene Watch Glasses > Diameter: 100mm; Transparent