Learn More
Kartell™ Transparent Polypropylene Powder Funnels
BIOPHARMA
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
Brand:  Kartell™ 0016700
Product Code. 12539746
  • 728.00 NOK / Pack of 20
Only null left
Estimated Shipment: 08-07-2025
Add to basket
Description
Description
Manufactured with polypropylene for strength and durability. Kartell™ Transparent Polypropylene Powder Funnels are designed to funnel large quantities of powders or other solid or liquid materials without flow rate restrictions, powder segregation or bridging.
  • 60° angled sides
  • External ribbing avoids pressure lock
  • Can be used with liquids
  • Autoclavable
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT23S00447 Certificate TypeDeclaration of Conformity Date28/11/2024 Catalog Number0016700
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Transparent Polypropylene Powder Funnels > Capacity: 94mL; Length: 22mm