Learn More
BRAND™ Translucent Polypropylene Y-shape Tubing Connector
Constructed of translucent polypropylene. Accommodates three tubes with Y-shaped stem and arms.
300.00 NOK - 510.00 NOK
Specifications
Product Type | Tubing Connector |
---|---|
Material | Polypropylene |
Shape | Y |
Color | Translucent |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
12110062
|
BRAND™
152903 |
4 to 5 mm tubing I.D. |
369.00 NOK
Pack of 20 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
15271509
|
BRAND™
152905 |
6 to 7 mm tubing I.D. |
415.00 NOK
Pack of 20 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11833930
|
BRAND™
152900 |
3 mm tubing I.D. |
300.00 NOK
Pack of 20 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
11843970
|
BRAND™
152907 |
8 to 9 mm tubing I.D. |
484.00 NOK
Pack of 20 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10579461
|
BRAND™
152910 |
10 to 11 mm tubing I.D. |
510.00 NOK
Pack of 20 |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Description
- Translucent polypropylene construction
- Accommodates 3 tubes of the same diameter
Specifications
Tubing Connector | |
Polypropylene | |
Y | |
Translucent |