missing translation for 'onlineSavingsMsg'
Learn More
trans-4-Ethylcyclohexanecarboxylic acid, 98%, Thermo Scientific™ Product Code.: 11464967

trans-4-Ethylcyclohexanecarboxylic acid, 98%, Thermo Scientific™

Product Code. p-7026094
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11464967

Brand: Thermo Scientific Alfa Aesar B21897.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6833-47-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD01076323,MFCD01568857
InChI Key UNROFSAOTBVBBT-UHFFFAOYSA-N
Synonym trans-4-ethylcyclohexanecarboxylic acid, 4-ethylcyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4-ethyl, 4-trans-ethyl cyclohexane carboxylic acid, cyclohexanecarboxylic acid, 4-ethyl-, trans, 1s,4r-4-ethylcyclohexane-1-carboxylic acid, pubchem2491, acmc-209o2z, ksc352s1t, ksc352s2bShow MoreShow Less
PubChem CID 1490288
IUPAC Name 4-ethylcyclohexane-1-carboxylic acid
SMILES CCC1CCC(CC1)C(O)=O

Specifications

Melting Point 48°C to 51°C
Quantity 1 g
Formula Weight 156.23
Percent Purity 98%
Chemical Name or Material trans-4-Ethylcyclohexanecarboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH12T029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10140767 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10231133 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10145748 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10149781 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-Ethylcyclohexanecarboxylic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.