missing translation for 'onlineSavingsMsg'
Learn More

trans-4-(Boc-aminomethyl)cyclohexylamine, 97%, Thermo Scientific Chemicals

Product Code. 15439078
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192323-07-2
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD06657670,MFCD12408630,MFCD22395596
InChI Key HMMYZMWDTDJTRR-UHFFFAOYSA-N
Synonym cis-4-boc-aminomethyl cyclohexylamine, tert-butyl trans-4-aminocyclohexylmethylcarbamate, tert-butyl trans-4-aminocyclohexyl methyl carbamate, tert-butyl cis-4-aminocyclohexyl methyl carbamate, trans-4-boc-aminomethyl cyclohexylamine, 4-boc-aminomethyl-cyclohexylamine, tert-butyl n-4-aminocyclohexyl methyl carbamate, tert-butyl 4-aminocyclohexyl methyl carbamate, tert-butyl trans-4-aminocyclohexyl methylcarbamate
PubChem CID 21882540
IUPAC Name tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(N)CC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'