missing translation for 'onlineSavingsMsg'
Learn More
trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals Product Code.: 11418526

trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals

Product Code. 11418526
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11418526

Brand: Thermo Scientific Alfa Aesar A15960.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18409-17-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00014057
InChI Key AYQPVPFZWIQERS-VOTSOKGWSA-N
Synonym trans-2-octen-1-ol, e-oct-2-en-1-ol, e-2-octen-1-ol, 2-octenol, 2-octen-1-ol, e, 2-octen-1-ol, 2e-octen-1-ol, unii-k54st7v77u, 2-octen-1-ol, 2e, 2e-oct-2-en-1-olShow MoreShow Less
PubChem CID 5318599
IUPAC Name (E)-oct-2-en-1-ol
SMILES CCCCCC=CCO

Specifications

Density 0.85
Boiling Point 88°C (11 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.4465
Quantity 5 g
Beilstein 1720707
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material trans-2-Octen-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10144531 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberA15960.06,A15960.14,A15960.22
Lot Number10245538 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA15960.06,A15960.14,A15960.22
Lot Number10236066 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15960.06,A15960.14,A15960.22
Lot Number10144531 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15960.06,A15960.14,A15960.22
Lot NumberB6026B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15960.06,A15960.14,A15960.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.