Learn More
Thermo Scientific™ TraceGOLD TG-ALC Plus GC Column
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific™ TraceGOLD™ TG-ALC GC Column.
Brand:  Thermo Scientific™ 26063-3390
8772.98 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15274533
  • 8820.00 NOK / Each
Only null left
Estimated Shipment: 04-07-2025
Description
Employ the most widely used MS phase in gas chromatography with the 5% phenyl phase Thermo Scientific™ TraceGOLD™ TG-ALC GC Column.
  • Low polarity phase, 5% diphenyl/95% dimethyl polysiloxane
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity
  • Exceptional inertness ideal for analysis of active compounds
TRUSTED_SUSTAINABILITY
Specifications
Specifications
260°C
0.32 mm
1.8 ÎĽm
30 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ TraceGOLD TG-ALC Plus GC Column > 0.32mm I.D.