missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trace Amine Receptor 1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93938-0.02ml
This item is not returnable.
View return policy
Description
Trace Amine Receptor 1 Polyclonal antibody specifically detects Trace Amine Receptor 1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Trace Amine Receptor 1 | |
Polyclonal | |
Western Blot 1:500-1:2000 | |
MGC126874, MGC138399, TA1RP11-295F4.9, TaR-1, TAR1trace amine-associated receptor 1, trace amine associated receptor 1, Trace amine receptor 1taR-1, TRAR1 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAAR1 (NP_612200.1). TSTDIMLSSASIFHLSFISIDRYYAVCDPLRYKAKMNILVICVMIFISWSVPAVFAFGMIFLELNFKGAEEIYYKHVHCRGGCSVFFSKISGVLTFMTSFY | |
0.02 mL | |
GPCR | |
134864 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
PBS (pH 7.3), 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trace Amine Receptor 1 Antibody - Azide and BSA Free, Novus Biologicals™ > 0.02 mL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction