Learn More
IKA™ TR 2 Tube Insert
Brand: IKA™ 0004525200
This item is not returnable.
View return policy
Description
Specifications
TR 2 Tube Insert | |
IKA Trayster Basic and Trayster Digital, Suitable 24 x 5mL Tubes with 12mm dia. | |
1 Ea. |
Product Content Correction
Product Title
IKA™ TR 2 Tube Insert > For use With: IKA Trayster Basic and Trayster Digital, Suitable 24 x 5mL Tubes with 12mm dia.