Learn More
Ansell™ TouchNTuff™ Disposable Blue Nitrile Gloves
Innovative nitrile compound offers outstanding chemical and mechanical properties, comfort and dexterity. Ansell TouchNTuff™ Disposable Blue Nitrile Gloves are lighter and cooler, yet safer and more durable.
271.00 NOK
Specifications
Certifications/Compliance | EN420, EN374/1,2,3, ASTM D 6319, FDA 21 CFR 177-2600,Arrete 09/11/04, European Regulation 1935/2004,German Recommendation for Food Commodities. |
---|---|
Color | Blue |
Disposable | Single-use |
Laboratory/Cleanroom Use | Yes |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
11726843
|
Ansell™
92670100 |
10 |
271.00 NOK
Pack of 100 |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
11766803
|
Ansell™
92670070 |
7 |
271.00 NOK
Pack of 100 |
In Stock
Log in to see stock. |
||||||
11776803
|
Ansell™
92670080 |
8 |
271.00 NOK
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||||
11786803
|
Ansell™
92670090 |
9 |
271.00 NOK
Pack of 100 |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
Description
- Three times the puncture resistance of latex or PVC disposable gloves
- 100% nitrile; no waxes, silicone or plasticizers
- Powder-free
- No risk of contaminating food with corn starch used to powder gloves
- Textured finish on fingertips
- Improved grip
- Rolled cuff
- Provides added protection at wrist and ensures a secure fit
Specifications
EN420, EN374/1,2,3, ASTM D 6319, FDA 21 CFR 177-2600,Arrete 09/11/04, European Regulation 1935/2004,German Recommendation for Food Commodities. | |
Single-use | |
Yes | |
Powder Free | |
Rolled | |
240 mm |
Blue | |
Yes | |
Nitrile | |
0.11 mm | |
92-670 |
Product Title
Ansell™ TouchNTuff™ Disposable Blue Nitrile Gloves