Learn More
Unity Lab Services™ Total Care Warranty for Thermo Scientific Water Purification Systems
Total Care Warranty provides an upgrade to factory warranty. Includes 2 day response time (within specified location and criteria), priority technical support, and one preventative maintenance visit.
Brand: Unity Lab Services™ TCWTYWPS
16392.60 NOK valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Description
- Upgrade a basic factory warranty with priority Technical Support response, and faster on-site scheduling along with an annual preventative maintenance visit
- 2 business day response time (may vary by location) for corrective maintenance
- Includes parts and labor covered under factory warranty for corrective maintenance
- One scheduled Preventative Maintenance visit, including a preventative maintenance kit (where applicable)
- Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools.
- Access to online service knowledge base
- Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the terms and conditions
- Instrument Software/ Firmware updates included as needed to support a preventative maintenance or corrective maintenance visit
- Upgrades to new versions of software are not included
- 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
Specifications
Water Purification Systems | |
Priority Technical Support, and One Preventative Maintenance Visit | |
Total Care Warranty | |
Upgrades to New Software Versions |
Upgrade to Factory Warranty | |
2 Day Response Time (Within Specified Location and Criteria), Priority Technical Support, and One Preventative Maintenance Visit | |
2 Days (Within Specified Location and Criteria) |
Promotions
-
Multiple Manufacturers
Exclusive Promotion on Equipment and Instruments
Take advantage of our limited-time offer on top-quality equipment and instruments to elevate your research and laboratory capabilities.Promo Code: 24069Expires: 2025-06-30