Learn More
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm Product Code.: 15462227
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm
Product Code. 15462227
1 Ea., Cardboard Box, Each
Click to view available options
Quantity:
1 Ea.
Packaging:
Cardboard Box
Unit Size:
Each
Product Code. 15462227
Brand: Thermo Scientific Alfa Aesar 045365.KS
Request Bulk or Custom Format
CAS: 7782-42-5 | C | 12.01 g/mol
Toray Carbon Paper, PTFE treated, TGP-H-60 is used as a gas diffusion layer in fuel cell applications. It is suitable for use as a catalyst backing layer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Toray Carbon Paper
CAS 7782-42-5
Packaging Cardboard Box
Molecular Formula C
MDL Number MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731
Quantity 1 Ea.
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
Solubility Information Insoluble in water.
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES [C]
IUPAC Name carbon
Molecular Weight (g/mol) 12.01
PubChem CID 5462310
ChEBI CHEBI:27594
Product Title
Toray Carbon Paper, PTFE treated, TGP-H-60, 19x19cm >