missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Topoisomerase III alpha Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1600.00 NOK - 3250.00 NOK
Specifications
Antigen | Topoisomerase III alpha |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18398176
|
Bio-Techne
NBP3-15507-20UL |
20 μg |
1600.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329223
|
Bio-Techne
NBP3-15507-100UL |
100 μg |
3250.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Topoisomerase III alpha Polyclonal antibody specifically detects Topoisomerase III alpha in Mouse samples. It is validated for Western BlotSpecifications
Topoisomerase III alpha | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
DNA topoisomerase 3-alpha, TOP3, topoisomerase (DNA) III alpha | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Topoisomerase III alpha (XP_011522303.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology | |
PBS, 50% glycerol, pH7.3 | |
7156 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title