missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TOMM5 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16844175
|
Proteintech
25607-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16834175
|
Proteintech
25607-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOMM5, also named as C9orf105 and TOM5, has three isoforms with MW 6 kDa and 9-10 kDa.Specifications
TOMM5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
B1AXP6, Q8N4H5 | |
401505, 68512 | |
TOMM5 Fusion Protein Ag22275 | |
Primary | |
-20°C | |
TOMM5 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C9orf105, RP11 263I4.1, Tom5, TOMM5 | |
TOMM5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |