Learn More
Solveco Toluene 99% Solvent
Toluene is a solvent used in industrial or institutional applications, with uses including rubber, resins, fats, printing inks, paints, and varnishes.
Brand: Solveco 1225
Description
Toluene is a high-purity, aromatic hydrocarbon solvent widely recognized for its effectiveness in dissolving a variety of substances. Its versatile chemical properties make it an essential component in numerous industrial processes and institutional applications.
Specifications
Methylbenzene | |
110°C to 111°C | |
HDPE Container | |
99.90% |
108-88-3 | |
25 L | |
92.15 | |
Technical |
Safety and Handling
- 25L Toluene 99%
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P314-Get medical advice/attention if you feel unwell.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : toluene
Your input is important to us. Please complete this form to provide feedback related to the content on this product.