Learn More
TOC Standard, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
35 mL
Unit Size:
35mL
Specifications
Specifications
Quantity | 35 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
TOCK1825F1 | Certificate of Analysis | 12/06/2025 | RTOCK18 |
TOCK1825B1 | Certificate of Analysis | 12/03/2025 | RTOCK18 |
TOCK1824H1 | Certificate of Analysis | 14/11/2024 | RTOCK18 |
Product Content Correction
Product Title
TOC Standard, Reagecon™ >