missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | TNS3 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899534
|
Proteintech
20053-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889534
|
Proteintech
20053-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody recognizes all isoforms of TNS3.
The Tensin (Tns) family of proteins is involved in the maintece of cellular structure by anchoring actin filaments at the focal adhesion via F-actin binding and capping activities. Tns proteins also contain a Src homology 2 (SH2) domain and have the ability to be phosphorylated, suggesting a role in signal transduction cascades. These diverse characteristics indicate that Tns proteins may be important links between the cytoskeleton and signal transduction pathways. Tns3, also known as TEM6 or TENS1, localizes to the focal adhesions of the plasma membrane. It is predomitly expressed in thyroid and placenta but can also be found in heart, liver, brain, prostate, pancreas, kidney, lung, skeletal muscle and white blood cells. Tns3 is essential for proper growth and development, as suggested by growth retardation and death in a number of Tns3-/- mice.Specifications
TNS3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5SSZ5, Q68CZ2 | |
319939, 360980, 64759 | |
Peptide | |
Primary | |
-20°C | |
TNS3 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
H_NH049I23.2, TEM6, TENS1, tensin 3, TNS3, TPP, Tumor endothelial marker 6 | |
Tns3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title