missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1660.20 NOK - 4731.57 NOK
Specifications
Antigen | TNFSF14 |
---|---|
Concentration | 0.21 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifications
TNFSF14 | |
ELISA | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O43557, Q9QYH9 | |
301133, 50930, 8740 | |
TNFSF14 Fusion Protein Ag5380 | |
Primary | |
-20°C | |
TNFSF14 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CD258, HVEM L, HVEML, LIGHT, LTg, TNFSF14, TR2 | |
Tnfsf14 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TNFSF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech