missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP3 Mouse anti-Human, Rat, Clone: 3A11G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66695-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene was identified as a gene whose expression is rapidly induced by the tumor necrosis factor (TNF). The protein encoded by this gene is a zinc finger protein, and has been shown to inhibit NF-kappa B activation as well as TNF-mediated apoptosis. Knockout studies of a similar gene in mice suggested that this gene is critical for limiting inflammation by terminating TNF-induced NF-kappa B responses.Specifications
TNFAIP3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P21580 | |
TNFAIP3 | |
TNFAIP3 Fusion Protein Ag18150 | |
20 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
3A11G6 | |
Unconjugated | |
TNFAIP3 | |
A20, OTUD7C, TNF alpha induced protein 3, TNFA1P2, TNFAIP3, Zinc finger protein A20 | |
Mouse | |
Protein G | |
RUO | |
683206, 7128 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction