missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium ICP Standard, 10000 mg/L Ti in HNO3 5%, HF 2%, Honeywell Fluka™
Brand: Honeywell Fluka E1257-100ML
1546.13 NOK valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Titanium ICP Standard | |
100 mL | |
Standards for ICP and ICP-MS | |
<2 |