Learn More
Lovibond™ Tintometer™ Acid Concentration Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 505420
This item is not returnable.
View return policy
Description
- Acid Concentration Tablets
- Cleaning Acid Strength
- Bottle of 100
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Acid Concentration Tablets (Cleaning Acid Strength) |