missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin (II) chloride dihydrate, ExpertQ, Scharlab
Click to view available options
Quantity:
250 g
Specifications
Specifications
CAS | 10025-69-1 |
Quantity | 250 g |
Molecular Formula | Cl2H4O2Sn |
InChI Key | FWPIDFUJEMBDLS-UHFFFAOYSA-L |
SMILES | O.O.[Cl-].[Cl-].[Sn++] |
IUPAC Name | λ²-tin(2+) dihydrate dichloride |
Molecular Weight (g/mol) | 225.64 |
Grade | ExpertQ |
Safety and Handling
Product Identifier
- Tin(II) chloride dihydrate, reagent grade, ACS, ISO, Reag. Ph Eur
- Warning
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'