Learn More
Velp Scientifica™ Tin Foil Cups
Velp Scientifica™ Tin Foil Cups for use with Dumas Nitrogen Analyzer NDA 701 and 702.
Brand:  Velp Scientifica™ A00000153
222.28 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15329574
  • 229.00 NOK / Pack of 100
Only null left
Estimated Shipment: 26-08-2025
Description
  • For use with NDA 701 and 702
Specifications
Specifications
Tin Foil Cups
Dumas Nitrogen Analyzer
0.1 kg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Velp Scientifica™ Tin Foil Cups > For Use With: NDA 701 and 702