missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tildrakizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA541986
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Tildrakizumab is a high-affinity, humanized, IgG1 kappa antibody targeting interleukin 23 p19 that shows promise in the evolution of treatment strategy in chronic plaque psoriasis 3. The Food and pharmaceutical Administration (FDA) approved ILUMYA (tildrakizumab-asmn) for the treatment of adults with moderate-to-severe plaque psoriasis who are candidates for systemic therapy or phototherapy in March 2018. The approved recommended dosage of ILUMYA is a subcutaneous injection of 100 mg at Weeks 0, 4, and every 12 weeks thereafter 7. A study was performed on the pharmacokinetics of this pharmaceutical on various ethnicities. The pharmacokinetics of tildrakizumab were similar in Japanese, Caucasian, and Chinese subjects 5.Specifications
Tildrakizumab | |
Recombinant Monoclonal | |
Unconjugated | |
SCH900222; MK-3222 | |
Human IL23A | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1.06 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction