missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombospondin 1 Monoclonal Antibody (C6.7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513390
This item is not returnable.
View return policy
Description
MA5-13390 targets Thrombospondin in FACS, IF, IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, Ovine, Porcine, and Rat samples. The MA5-13390 immunogen is ca2+-replete, native purified human TSP from supernatant of thrombin-activated platelets.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.Specifications
Thrombospondin 1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P07996, Q28178 | |
THBS1 | |
Ca2+-replete, native purified human TSP from supernatant of thrombin-activated platelets | |
500 μL | |
Primary | |
Bovine, Canine, Equine, Human, Ovine, Porcine, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
C6.7 | |
Unconjugated | |
THBS1 | |
glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
Mouse | |
Protein G | |
RUO | |
100057478, 101106433, 281530, 445442, 487486, 492313, 7057 | |
4° C | |
Liquid |