Learn More
SCAT™ Polypropylene Thread Adapters
Safety Caps and Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
Brand:  SCAT™ 107092
597.49 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10170130
  • 710.00 NOK / Each
Only null left
Estimated Shipment: 21-07-2025
Description
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polypropylene (PP) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Specifications
Specifications
Thread 1: S51 (m), Thread S55 (f)
Colorless
Polypropylene
Thread Adapter
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SCAT™ Polypropylene Thread Adapters > Thread Adapters; Closure Size: Thread 1: S51 (m), Thread S55 (f); Color: Colorless