Learn More
Bohlender™ Thermocoupler
Brand:  Bohlender™ P1730-25
3037.56 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10552973
  • 3255.00 NOK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
Thermocouple
500 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bohlender™ Thermocoupler > Length: 500mm