Learn More
Thermo Scientific™ Valco Accessories
Valco continues to offer a diverse line in terms of number of ports, fitting sizes, and materials of construction.
Brand: Thermo Scientific™ ZF1-10
581.20 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Ferrule | |
Stainless Steel | |
0.062 in. | |
0.15 cm |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Product Title
Thermo Scientific™ Valco Accessories > 1/16in. SS ferrules