Learn More
Epredia™ Shandon™ Plastic Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Brand:  Epredia™ 72110013
9500.06 NOK valid until 2025-08-31
Use promo code "24157" to get your promotional price.
 View more versions of this product
Product Code. 11927774
  • 10990.00 NOK / Pack of 250
Spotlight
Use promo code 24157 at checkout
View Available Promotions
Description
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Coverplate
250/Cs.
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Save big on essential products for cancer research
    Promo Code24147
    Expires2025-07-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Epredia™ Shandon™ Plastic Coverplates > Coverplate; 250/Cs.