Learn More
Thermo Scientific™ Septa
Brand: Thermo Scientific™ 31303215-BP
1218.70 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Water analysis
File size: 7.3 MB
Product Content Correction
Product Title
Thermo Scientific™ Septa > Auto Sep T Septa; 9.5mm; Pack of 25