Learn More
Thermo Scientific™ Nalgene™ Petri Dish
Shatterproof and reusable
Brand: Thermo Scientific™ 5500-0010
4422.14 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Resists cell adhering
- Stackable for convenience
- Top and bottom interlocking ridges
- Autoclavable
Specifications
Autoclavable | |
Round | |
Petri Dish | |
Pk, of 10 | |
100 mm |
PMP | |
Sterile | |
Stackable | |
15 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1424887 | Certificate of Compliance | 13/03/2025 | 5500-0010 |
1424886 | Certificate of Compliance | 13/03/2025 | 5500-0010 |
1419588 | Certificate of Compliance | 13/12/2024 | 5500-0010 |
1290462 | Certificate of Compliance | 10/12/2024 | 5500-0010 |
1321913 | Certificate of Compliance | 10/12/2024 | 5500-0010 |
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Petri Dish > 4 x .5 in. (100 x 15mm)