Learn More
Thermo Scientific™ Forma™ Chamber Cooling Coil
For Use With: Forma Series II Water jacketed CO2 Incubator
Brand: Thermo Scientific™ 190645
18417.13 NOK valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
- Precise control
- Choice of TC or IR sensors
- Superior parameter recovery characteristics
- Continuous contamination control technology
Specifications
TC Sensor or IR Sensor | |
Forma Series II Water jacketed CO2 incubator | |
LED | |
Chamber Cooling Coil |
Product Content Correction
Product Title
Thermo Scientific™ Forma™ Chamber Cooling Coil > Chamber Cooling Coil; use with refrigerated water bath/circulator to operate incubator at lower than ambient temperatures; Factory installed