Learn More
Thermo Scientific™ Clear Strong Heat Sealing Film
Transparent—suitable for genotyping applications
Brand: Thermo Scientific™ AB-0685
2265.35 NOK valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
- Clear polyester polypropylene laminate heat seal
- Strong and unpeelable
- Also suitable for storage and disposal of hazardous material
- Good solvent resistance; recommended for use with DMSO
- Seal integrity range: -80°C to +120°C
Specifications
Clear | |
Non-sterile |
Promotions
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30