Learn More
Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
5808.90 NOK - 8829.71 NOK
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Includes | Price | Quantity & Availability | |||||
11880933
|
Thermo Scientific™
2122 |
1 L | Carrying Handle, Vented Secure Lid |
6236.06 NOK
Each |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
11868003
|
Thermo Scientific™
2123 |
2 L | Carrying Handle, Vented Secure Lid |
6442.98 NOK
Each |
In Stock
Log in to see stock. ![]() |
|||||
11878003
|
Thermo Scientific™
2124 |
4.5 L | Carrying Handle, Vented Secure Lid |
8829.71 NOK
Each |
In Stock
Log in to see stock. |
|||||
13488119
|
Thermo Scientific™
2129 |
1 L | Without Lid and Handle |
6254.87 NOK
Each |
In Stock
Log in to see stock. |
|||||
13478129
|
Thermo Scientific™
2130 |
1.9 L | Without Lid and Handle |
5808.90 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
Stainless Steel:- Durable, corrosion-resistant stainless-steel exterior
- Carrying handle and vented secure lid (Part numbers 2122, 2123, 2124)
Shallow Wide Mouth:
- Ideal for use as a cold trap
- Convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol
- Stainless-steel jacket and cushioned base for years of trouble-free use
- Without lid and handle (Part number 2129)
- Warranty varies by country, please contact us for details
Specifications
Stainless Steel | |
Benchtop |