Learn More
TFA Dostmann™ Thermo Barometer
Brand:  TFA Dostmann™ 45.1000.05
Product Code. 11848272
  • 387.00 NOK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Specifications
Beech
121 mm
Product Title
TFA Dostmann™ Thermo Barometer > Diameter: 121mm