Learn More
LTE Scientific™ Thermal Data Printer
Thermal Data Printer for LTE Touchlave models.
Brand:  LTE Scientific™ TC/TDP/02
 View more versions of this product
Product Code. 12735405
  • 13920.00 NOK / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Thermal data printer
  • Prints all cycle data as cycle progresses
  • Printout includes any faults/alerts
  • Provides printed validation of load success
Specifications
Specifications
All LTE Touchclave R Model Autoclaves
TC/TDP/02
Thermal Data Printer
Non-sterile