Learn More
Abeba™ The Original 5080 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
636.00 NOK
Specifications
Certifications/Compliance | CE EN ISO 20347:2012 OB A, FO SRC/li |
---|---|
Color | White |
Material | Leather |
Sole Features | Slip-resistant |
Style | 5080 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15306666
|
Abeba™
5080/35 |
35 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15316666
|
Abeba™
5080/36 |
36 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15326666
|
Abeba™
5080/37 |
37 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15336666
|
Abeba™
5080/38 |
38 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15346666
|
Abeba™
5080/39 |
39 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15356666
|
Abeba™
5080/40 |
40 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15366666
|
Abeba™
5080/41 |
41 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15376666
|
Abeba™
5080/42 |
42 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
15386666
|
Abeba™
5080/43 |
43 |
636.00 NOK
1 pair |
Estimated Shipment: 10-02-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20347:2012 OB A, FO SRC
- Smooth leather, perforated
- White/mint
- Slip resistance outsole sole
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
- Leather top layer
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li | |
Leather | |
5080 |
White | |
Slip-resistant | |
Animal fats, oil and fuel |