Learn More
TH1L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28417-1-AP-20UL
This item is not returnable.
View return policy
Description
The TH1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TH1L. This antibody recognizes human antigen. The TH1L antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
TH1L | |
Polyclonal | |
Unconjugated | |
20 μL | |
51497 | |
IgG |
Western Blot, Immunofluorescence, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |