Learn More
TH1L Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28417-1-AP-150UL
 View more versions of this product
Product Code. 16836135
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
The TH1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TH1L. This antibody recognizes human antigen. The TH1L antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
TH1L
Polyclonal
Unconjugated
150 μL
51497
IgG
Western Blot, Immunofluorescence, ELISA
333 μg/mL