Learn More
Texwipe™ Direct Thermal Cleanroom Labels
Designed for critical environments where direct thermal printing is used
Brand:  Texwipe™ TX532D
 View more versions of this product
Product Code. 15501255
  • 13005.00 NOK / 4 rolls
Only null left
Estimated Shipment: 12-09-2025
Description
  • Low out-gassing polypropylene made with ultra-low leachables and low particulates
  • Quick-ship pressure sensitive
  • Available with two different adhesive levels providing permanent adhesion without leaving residue
  • Tested for surface particles, cleaned and double bagged
Specifications
Specifications
Acrylic
Polypropylene
1000/Pk.
7.62 x 5 cm
-29 to 60°C
Label
Designed for critical environments where direct thermal printing is used
3 x 2 in.
-20 to 140°F
3.3 mil
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Texwipe™ Direct Thermal Cleanroom Labels > 3 x 2 in.