Learn More
Tetrahydro-2,2-dimethyl-4H-pyran-4-one, 95%, Thermo Scientific Chemicals
876.00 NOK - 3805.00 NOK
Chemical Identifiers
CAS | 1194-16-7 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD01549337 |
InChI Key | BWMNOXJVRHGUQM-UHFFFAOYSA-N |
Synonym | 2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l |
PubChem CID | 1738159 |
IUPAC Name | 2,2-dimethyloxan-4-one |
SMILES | CC1(C)CC(=O)CCO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15473777
|
Thermo Scientific Alfa Aesar
H27774.MD |
250 mg | N/A | ||||||
11362949
|
Thermo Scientific Alfa Aesar
H27774.03 |
1 g |
3805.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly soluble in water.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Chemical Identifiers
1194-16-7 | |
128.17 | |
BWMNOXJVRHGUQM-UHFFFAOYSA-N | |
1738159 | |
CC1(C)CC(=O)CCO1 |
C7H12O2 | |
MFCD01549337 | |
2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l | |
2,2-dimethyloxan-4-one |
Specifications
1194-16-7 | |
C7H12O2 | |
2,2-dimethyltetrahydropyran-4-one, 2,2-dimethyldihydro-2h-pyran-4 3h-one, 2,2-dimethyl-tetrahydro-pyran-4-one, tetrahydro-2,2-dimethyl-4h-pyran-4-one, 2,2-dimethyltetrahydro-4h-pyran-4-one, 4h-pyran-4-one, tetrahydro-2,2-dimethyl, 2,2-dimethyl-2h-3,5,6-trihydropyran-4-one, pubchem24423, acmc-1bt7l | |
BWMNOXJVRHGUQM-UHFFFAOYSA-N | |
2,2-dimethyloxan-4-one | |
1738159 | |
95% |
60°C to 64°C (8 mmHg) | |
MFCD01549337 | |
Slightly soluble in water. | |
CC1(C)CC(=O)CCO1 | |
128.17 | |
128.17 | |
Tetrahydro-2,2-dimethyl-4H-pyran-4-one |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold