Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, with Beaded Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
284.00 NOK - 518.00 NOK
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15852285
|
Fisherbrand™
90106153 |
3 mL | 75 mm | 10 mm |
453.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17921152
|
Fisherbrand™
90106192 |
5 mL | 75 mm | 12 mm |
284.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17951132
|
Fisherbrand™
90106193 |
13 mL | 100 mm | 16 mm |
471.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17941132
|
Fisherbrand™
90106194 |
17 mL | 125 mm | 16 mm |
482.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17901152
|
Fisherbrand™
90106196 |
21 mL | 150 mm | 16 mm |
499.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17961122
|
Fisherbrand™
90106186 |
21 mL | 160 mm | 16 mm |
505.00 NOK
Pack of 100 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17931152
|
Fisherbrand™
90106197 |
27 mL | 150 mm | 18 mm |
518.00 NOK
Pack of 100 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||
17911152
|
Fisherbrand™
90106198 |
53 mL | 150 mm | 24 mm |
398.00 NOK
Pack of 50 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
17971122
|
Fisherbrand™
90106187 |
57 mL | 150 mm | 25 mm |
414.00 NOK
Pack of 50 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||
Description
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for this work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from borosilicate glass 3.3
- Thick-walled and therefore mechanically very resistant
- With round bottom
Product Title
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, with Beaded Rim, Medium Wall