Learn More
Velp Scientifica™ Test Tubes
Velp Scientifica™ Test Tubes are spare tubes for use with TMD 6 Mineralization System.
Brand: Velp Scientifica™ 10000000
638.75 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Spare test tube
- Features a spherical joint
Specifications
Test Tube | |
Spherical joint | |
42 mm | |
250 mL |
Product Content Correction
Product Title
Velp Scientifica™ Test Tubes > Diameter: 42 mm