Learn More
Kartell™ Plastilab™ Test Tube Caps
Fits plastic or glass test tubes.
Brand:  Kartell™ 0065600
 View more versions of this product
Product Code. 10316351
  • 154.00 NOK / Pack of 100
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Description
  • Made from polyethylene
Specifications
Specifications
CE Certified
28 mm
Test Tube Plug Cap
Documents
One moment while we fetch your results.
Product Title
Kartell™ Plastilab™ Test Tube Caps > Dia.: 28mm; For use with: 30 Dia. Plastic or Glass Tube