Learn More
tert-Butyldichlorophosphine, Thermo Scientific Chemicals Product Code.: 10195790
tert-Butyldichlorophosphine, Thermo Scientific Chemicals
Product Code. p-80246 Shop All Thermo Scientific Chemicals Products
2.5 g, Glass bottle, 2.50g
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.50g
Product Code. 10195790
Brand: Thermo Scientific Acros 300530025
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25979-07-1
Molecular Formula C4H9Cl2P
Molecular Weight (g/mol) 158.98
MDL Number MFCD00013615
InChI Key NMJASRUOIRRDSX-UHFFFAOYSA-N
Synonym tert-butyldichlorophosphine, t-butyldichlorophosphine, tert-c4h9pcl2, tert-butyl dichloro phosphane, phosphonous dichloride, 1,1-dimethylethyl, snpladsptpdwuup@, pubchem6471, acmc-1cmec, tert-butyl dichloro phosphine, tert-butylphosphine dichloride
PubChem CID 117690
IUPAC Name tert-butyl(dichloro)phosphane
SMILES CC(C)(C)P(Cl)Cl
Specifications
Melting Point 49.0°C
Color White to Yellow
Boiling Point 142.0°C to 150.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (CH3)3CPCl2
Quantity 2.5 g
Solubility Information Solubility in water: reacts. Other solubilities: soluble in n-pentane,n-hexane,ether,toluene,,thf
Formula Weight 158.98
Physical Form Crystalline Low Melting Solid
Chemical Name or Material tert-Butyldichlorophosphine
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0458030 Certificate TypeCertificate of Analysis Date23/11/2023 Product Code300530100,300530025
Lot NumberA0429160 Certificate TypeCertificate of Analysis Date03/11/2023 Product Code300530100,300530025
Lot NumberA0413641 Certificate TypeCertificate of Analysis Date30/05/2022 Product Code300530100,300530025
Lot NumberA0399237 Certificate TypeCertificate of Analysis Date11/02/2021 Product Code300530100,300530025
Product Title
tert-Butyldichlorophosphine, Thermo Scientific Chemicals >