missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 15428946

tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15428946
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15428946

Brand: Thermo Scientific Alfa Aesar H64468.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77497-74-6
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
MDL Number MFCD04114841
InChI Key KRSUDBIZXKOKGA-LBPRGKRZSA-N
Synonym s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester, tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester, tic-obut, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac, tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochlorideShow MoreShow Less
PubChem CID 9209323
IUPAC Name tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
SMILES CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1

Specifications

Quantity 5 g
Beilstein 5744730
Solubility Information Soluble in water.
Formula Weight 269.77
Percent Purity 99%
Physical Form Solid
Chemical Name or Material tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81400055 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64468.03,H64468.MD,H64468.06
Lot Number5005F23Q Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64468.03,H64468.MD,H64468.06
Lot Number5011K27N Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64468.03,H64468.MD,H64468.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.