Learn More
Memmert™ Temperature Restriction for UN30
Optional temperature restriction between 60°C and 250°C.
Brand: Memmert™ UN30A8
This item is not returnable.
View return policy
Description
- Accessory
- 1 Choice
- Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
UN30 Oven | |
Temperature Restriction |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Memmert™ Temperature Restriction for UN30 > For Use With: UN30