Learn More
Testo™ Testoterm™ Temperature Strips
Use these strips for monitoring the temperature of moving or small objects and long-term monitoring of transport and storage temperatures. Testoterm™ Temperature Strips can be applied to a variety of different surfaces and react to specific increases in temperature by changing color irreversibly within 2 to 3 seconds.
417.82 NOK - 570.00 NOK
Specifications
Mounting | Self-adhesive |
---|---|
Accuracy (Temperature) | ±1.5°C |
Type | Self-adhesive Temperature Strips |
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | ||||||
10028660
|
Testo™
0646 0108 |
37°C to 65°C |
417.82 NOK
Pack of 10 |
Estimated Shipment: 08-07-2025
Log in to see stock. |
||||||
10282161
|
Testo™
0646 0916 |
71°C to 110°C |
570.00 NOK
Pack of 10 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
Description
- Self-adhesive temperature strips
- Available in booklets of 10
- Strips are peeled off and applied to the object you wish to measure
- Change color when thresholds are exceeded within 2 to 3 seconds
Specifications
Self-adhesive | |
±1.5°C | |
Self-adhesive Temperature Strips |