Learn More
OHAUS™ Temperature Probe
Hot plates and stirrers accessory
Brand: OHAUS™ 30500590
1605.16 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Temperature probe in stainless steel or PTFE
- Varying sizes available
Specifications
20 cm | |
Temperature Probe | |
Stainless Steel |
Product Content Correction
Product Title
OHAUS™ Temperature Probe > 20 cm, Stainless Steel